4-(Morpholin-4-ylmethyl)benzonitrile

Supplier: Matrix Scientific

Synonyms: 4-(Morpholinomethyl)benzonitrile

060411-500MG
101920-008EA 181.7 USD
101920-008
4-(Morpholin-4-ylmethyl)benzonitrile
4-(Morpholin-4-ylmethyl)benzonitrile

Hazard: Irritant

Formula: C₁₂H₁₄N₂O
MW: 202.2500 g/mol
MDL Number: MFCD00454271
CAS Number: 37812-51-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR