DL-Alanine methyl ester hydrochloride
Supplier: Matrix Scientific
Synonyms:
H-DL-Ala-OMe.HCl
060389-500MG
101919-966EA
183.08
USD
101919-966
DL-Alanine methyl ester hydrochloride
DL-Alanine methyl ester hydrochloride
CAS: 13515-97-4
MDL Number: MFCD00035523
Molecular Formula: C4H10ClNO2
Molecular Weight: 139,58
Hazard: Irritant
Formula:
C₄H₉NO₂ MW: 139.58 g/mol |
MDL Number:
MFCD00035523 CAS Number: 13515-97-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...