1-Methyl-5-(trifluoromethyl)-1H-benzimidazol-2-amine hydrobromide

Supplier: Matrix Scientific

060185-500MG
101919-574EA 243.08 USD
101919-574
1-Methyl-5-(trifluoromethyl)-1H-benzimidazol-2-amine hydrobromide
1-Methyl-5-(trifluoromethyl)-1H-benzimidazol-2-amine hydrobromide

Hazard: Irritant

Formula: C₉H₉BrF₃N₃
MDL Number: MFCD19442219
CAS Number: 945023-32-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR