2-(4,6-Dichloro-2-methylpyrimidin-5-yl)acetaldehyde ≥96%

Supplier: Matrix Scientific

059855-1G
101919-306EA 412.31 USD
101919-306
2-(4,6-Dichloro-2-methylpyrimidin-5-yl)acetaldehyde ≥96%
2-(4,6-Dichloro-2-methylpyrimidin-5-yl)acetaldehyde

Hazard: Irritant

Formula: C₇H₆Cl₂N₂O
CAS Number: 474656-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR