6-Nitro-4-quinazolone

Supplier: Matrix Scientific

Synonyms: 6-Nitroquinazolin-4(3H)-one6-Nitro-4(3H)-quinazolinone

059719-500MG
101918-948EA 49.23 USD
101918-948
6-Nitro-4-quinazolone
6-Nitro-4-quinazolone

Hazard: Irritant

Formula: C₈H₅N₃O₃
MW: 191.1436 g/mol
Melting Pt: 286…287 °C
MDL Number: MFCD00137845
CAS Number: 6943-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR