3-Fluoro-5-methoxybenzoic acid ≥98%

Supplier: Matrix Scientific

059599-5G 059599-1G
101918-614EA 32.31 USD
101918-614 101918-616
3-Fluoro-5-methoxybenzoic acid ≥98%
3-Fluoro-5-methoxybenzoic acid

Hazard: Irritant

Formula: C₈H₇FO₃
MW: 170.1400 g/mol
MDL Number: MFCD08235092
CAS Number: 176548-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR