1,4,5,6-Tetrahydrocyclopenta[c]pyrazole-3-carbohydrazide

Supplier: Matrix Scientific

054624-500MG
101907-948EA 164.62 USD
101907-948
1,4,5,6-Tetrahydrocyclopenta[c]pyrazole-3-carbohydrazide
1,4,5,6-Tetrahydrocyclopenta[c]pyrazole-3-carbohydrazide

Hazard: Irritant

Formula: C₇H₁₀N₄O
CAS Number: 299166-55-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR