1-[5-(2-Thienyl)-1H-pyrazol-3-yl]methanamine

Supplier: Matrix Scientific

054614-500MG
101907-928EA 306.15 USD
101907-928
1-[5-(2-Thienyl)-1H-pyrazol-3-yl]methanamine
1-[5-(2-Thienyl)-1H-pyrazol-3-yl]methanamine

Hazard: Irritant

Formula: C₈H₉N₃S
MDL Number: MFCD10036164
CAS Number: 859850-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR