2,4-Dichloro-3-fluorobenzoic acid ≥95%

054586-1G
101907-864EA 290.43 USD
101907-864
2,4-Dichloro-3-fluorobenzoic acid ≥95%
2,4-Dichloro-3-fluorobenzoic acid

Hazard: Irritant

Formula: C₇H₃Cl₂FO₂
MDL Number: MFCD14707646
CAS Number: 915145-05-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR