Ethyl-1-(3-aminopropyl)-3-piperidinecarboxylate ≥95%
Supplier: Matrix Scientific
054573-500MG
101907-818EA
556.92
USD
101907-818
Ethyl-1-(3-aminopropyl)-3-piperidinecarboxylate ≥95%
Ethyl-1-(3-aminopropyl)-3-piperidinecarboxylate
Hazard: Irritant
Formula:
C₁₁H₂₂N₂O₂ MW: 214.31 g/mol |
MDL Number:
MFCD14584771 CAS Number: 1221792-43-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...