5-Bromo-3-chloro-1H-indazole ≥95%

Supplier: Matrix Scientific

054369-500MG
101907-176EA 160 USD
101907-176
5-Bromo-3-chloro-1H-indazole ≥95%
5-Bromo-3-chloro-1H-indazole

Hazard: Irritant

Formula: C₇H₄BrClN₂
MW: 231.4800 g/mol
MDL Number: MFCD06738284
CAS Number: 36760-19-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR