6-Bromo-3-chloro-1H-indazole ≥95%

Supplier: Matrix Scientific

054368-500MG
101907-174EA 921.54 USD
101907-174
6-Bromo-3-chloro-1H-indazole ≥95%
6-Bromo-3-chloro-1H-indazole

Hazard: Irritant

Formula: C₇H₄BrClN₂
MW: 231.48 g/mol
MDL Number: MFCD06738283
CAS Number: 885271-78-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR