Diethyl-7-oxo-4,7-dihydropyrazolo[1,5-a]pyrimidine-3,6-dicarboxylate ≥95%

Supplier: Matrix Scientific

054357-500MG
101907-154EA 747.69 USD
101907-154
Diethyl-7-oxo-4,7-dihydropyrazolo[1,5-a]pyrimidine-3,6-dicarboxylate ≥95%
Diethyl-7-oxo-4,7-dihydropyrazolo[1,5-a]pyrimidine-3,6-dicarboxylate

Hazard: Irritant

Formula: C₁₂H₁₃N₃O₅
MDL Number: MFCD01812303
CAS Number: 155374-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR