2-(4-Bromoanilino)-1-phenyl-1-ethanol ≥95%

Supplier: Matrix Scientific

054320-500MG 054320-5G 054320-1G
101907-058EA 806.15 USD
101907-058 101907-054 101907-056
2-(4-Bromoanilino)-1-phenyl-1-ethanol ≥95%
2-(4-Bromoanilino)-1-phenyl-1-ethanol

Hazard: Irritant

Formula: C₁₄H₁₄BrNO
MDL Number: MFCD11959450
CAS Number: 91851-17-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR