2,4-Dichloro-5-(trifluoromethyl)pyridine ≥95%

Supplier: Matrix Scientific

054308-1G
101907-012EA 643.08 USD
101907-012
2,4-Dichloro-5-(trifluoromethyl)pyridine ≥95%
2,4-Dichloro-5-(trifluoromethyl)pyridine

Hazard: Irritant

Formula: C₆H₂Cl₂F₃N
MDL Number: MFCD14525480
CAS Number: 888327-38-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR