Ethyl-3-chloro-4-(trifluoromethyl)picolinate ≥95%
Supplier: Matrix Scientific
Synonyms:
Ethyl 3-chloro-4-(trifluoromethyl)pyridine-2-carboxylate
054292-1G
054292-500MG
101906-942EA
846.15
USD
101906-942
101906-944
Ethyl-3-chloro-4-(trifluoromethyl)picolinate ≥95%
Ethyl 3-chloro-4-(trifluoromethyl)picolinate
Hazard: Irritant
Formula:
C₉H₇ClF₃NO₂ |
MDL Number:
MFCD14581663 CAS Number: 1198475-44-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...