4-Iodooxindole ≥95%

Supplier: Matrix Scientific

054244-1G 054244-500MG
101906-744EA 533.85 USD
101906-744 101906-742
4-Iodooxindole ≥95%
4-Iodooxindole

Hazard: Irritant

Formula: C₈H₆INO
CAS Number: 179536-52-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR