Methyl-3-amino-5-bromo-2-hydroxybenzenecarboxylate ≥95%

Supplier: Matrix Scientific

054242-1G 054242-500MG
101906-736EA 927.69 USD
101906-736 101906-734
Methyl-3-amino-5-bromo-2-hydroxybenzenecarboxylate ≥95%
Methyl-3-amino-5-bromo-2-hydroxybenzenecarboxylate

Hazard: Irritant

Formula: C₈H₈BrNO₃
CAS Number: 141761-82-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR