1-Benzyl-4-(methylamino)-4-piperidinecarbonitrile ≥95%
Supplier: Matrix Scientific
054208-1G
054208-500MG
101906-588EA
698.46
USD
101906-588
101906-586
1-Benzyl-4-(methylamino)-4-piperidinecarbonitrile ≥95%
1-Benzyl-4-(methylamino)-4-piperidinecarbonitrile
Hazard: Irritant
Formula:
C₁₄H₁₉N₃ |
MDL Number:
MFCD00023748 CAS Number: 953-79-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...