1-(5-Amino-3-chloro-1H-indazol-1-yl)-1-ethanone ≥95%

Supplier: Matrix Scientific

054199-500MG
101906-548EA 1109.23 USD
101906-548
1-(5-Amino-3-chloro-1H-indazol-1-yl)-1-ethanone ≥95%
1-(5-Amino-3-chloro-1H-indazol-1-yl)-1-ethanone

Hazard: Irritant

Formula: C₉H₈ClN₃O
MDL Number: MFCD14560532
CAS Number: 1203897-92-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR