6-Hydroxy-2-(methylsulfanyl)-4-pyrimidinecarboxylic acid ≥95%

Supplier: Matrix Scientific

054192-500MG
101906-522EA 653.85 USD
101906-522
6-Hydroxy-2-(methylsulfanyl)-4-pyrimidinecarboxylic acid ≥95%
6-Hydroxy-2-(methylsulfanyl)-4-pyrimidinecarboxylic acid

Hazard: Irritant

Formula: C₆H₆N₂O₃S
MDL Number: MFCD00023236
CAS Number: 6314-14-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR