2,3-Dihydro-1H-pyrido[3,4-b][1,4]oxazine

Supplier: Matrix Scientific

054184-500MG 054184-100MG
101906-496EA 800 USD
101906-496 101906-494
2,3-Dihydro-1H-pyrido[3,4-b][1,4]oxazine
2,3-Dihydro-1H-pyrido[3,4-b][1,4]oxazine

Hazard: Irritant

Formula: C₇H₈N₂O
MDL Number: MFCD13176662
CAS Number: 194022-45-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR