1-(2-((tert-Butyldimethylsilyloxy)methyl)furo[3,2-b]pyridin-6-yl)ethanone
Supplier: Matrix Scientific
054181-500MG
054181-100MG
101906-484EA
800
USD
101906-484
101906-482
1-(2-((tert-Butyldimethylsilyloxy)methyl)furo[3,2-b]pyridin-6-yl)ethanone
1-(2-((tert-Butyldimethylsilyloxy)methyl)furo[3,2-b]pyridin-6-yl)ethanone
Hazard: Irritant
Formula:
C₁₆H₂₃NO₃Si |
MDL Number:
MFCD13563102 CAS Number: 1203499-37-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...