6-((3-((tert-Butyldimethylsilyloxy)methyl)pyrrolidin-1-yl)methyl)furo[3,2-b]pyridine

Supplier: Matrix Scientific

054179-500MG 054179-100MG
101906-476EA 800 USD
101906-476 101906-474
6-((3-((tert-Butyldimethylsilyloxy)methyl)pyrrolidin-1-yl)methyl)furo[3,2-b]pyridine
6-((3-((tert-Butyldimethylsilyloxy)methyl)pyrrolidin-1-yl)methyl)furo[3,2-b]pyridine

Hazard: Irritant

Formula: C₁₉H₃₀N₂O₂Si
MW: 346.55 g/mol
MDL Number: MFCD13176660
CAS Number: 1188994-30-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR