6-Bromofuro[3,2-b]pyridine-2-carbonitrile

Supplier: Matrix Scientific

054171-100MG
101906-442EA 244.62 USD
101906-442
6-Bromofuro[3,2-b]pyridine-2-carbonitrile
6-Bromofuro[3,2-b]pyridine-2-carbonitrile

Hazard: Irritant

Formula: C₈H₃BrN₂O
MDL Number: MFCD13563100
CAS Number: 1203498-94-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR