Furo[3,2-b]pyridine-6-carbonitrile

Supplier: Matrix Scientific

054169-500MG 054169-100MG
101906-436EA 775.38 USD
101906-436 101906-434
Furo[3,2-b]pyridine-6-carbonitrile
Furo[3,2-b]pyridine-6-carbonitrile

Hazard: Irritant

Formula: C₈H₄N₂O
MDL Number: MFCD13563098
CAS Number: 1203499-65-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR