tert-Butyl-3-(3-hydroxyprop-1-ynyl)-5-methyl-1H-pyrrolo[2,3-b]pyridine-1-carboxylate, Technical Grade
Supplier: Matrix Scientific
054166-100MG
054166-500MG
101906-424EA
800
USD
101906-424
101906-422
tert-Butyl-3-(3-hydroxyprop-1-ynyl)-5-methyl-1H-pyrrolo[2,3-b]pyridine-1-carboxylate, Technical Grade
tert-Butyl-3-(3-hydroxyprop-1-ynyl)-5-methyl-1H-pyrrolo[2,3-b]pyridine-1-carboxylate
Hazard: Irritant
Formula:
C₁₆H₁₈N₂O₃ MW: 286.33 g/mol |
MDL Number:
MFCD13563095 CAS Number: 1198106-25-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...