tert-Butyl-3-(hydroxymethyl)-5-methyl-1H-pyrrolo[2,3-b]pyridine-1-carboxylate

Supplier: Matrix Scientific

054164-100MG 054164-500MG
101906-414EA 364.62 USD
101906-414 101906-416
tert-Butyl-3-(hydroxymethyl)-5-methyl-1H-pyrrolo[2,3-b]pyridine-1-carboxylate
tert-Butyl-3-(hydroxymethyl)-5-methyl-1H-pyrrolo[2,3-b]pyridine-1-carboxylate

Hazard: Irritant

Formula: C₁₄H₁₈N₂O₃
MW: 262.31 g/mol
MDL Number: MFCD13563093
CAS Number: 1198103-73-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR