tert-Butyl-3-iodo-5-methyl-1H-pyrrolo[2,3-b]pyridine-1-carboxylate

Supplier: Matrix Scientific

054156-100MG 054156-500MG
101906-384EA 800 USD
101906-384 101906-382
tert-Butyl-3-iodo-5-methyl-1H-pyrrolo[2,3-b]pyridine-1-carboxylate
tert-Butyl-3-iodo-5-methyl-1H-pyrrolo[2,3-b]pyridine-1-carboxylate

Hazard: Irritant

Formula: C₁₃H₁₅IN₂O₂
MW: 358.18 g/mol
MDL Number: MFCD13176648
CAS Number: 1198097-29-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR