7-Chloro-5-iodo-1-(triisopropylsilyl)-1H-pyrrolo[2,3-c]pyridine
Supplier: Matrix Scientific
054153-100MG
054153-500MG
101906-372EA
800
USD
101906-372
101906-370
7-Chloro-5-iodo-1-(triisopropylsilyl)-1H-pyrrolo[2,3-c]pyridine
7-Chloro-5-iodo-1-(triisopropylsilyl)-1H-pyrrolo[2,3-c]pyridine
Hazard: Irritant
Formula:
C₁₆H₂₄ClIN₂Si |
MDL Number:
MFCD13176646 CAS Number: 1198097-05-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...