5-Methyl-1H-pyrrolo[2,3-b]pyridine-3-carbaldehyde
Supplier: Matrix Scientific
054145-100MG
054145-500MG
101906-340EA
800
USD
101906-340
101906-338
5-Methyl-1H-pyrrolo[2,3-b]pyridine-3-carbaldehyde
5-Methyl-1H-pyrrolo[2,3-b]pyridine-3-carbaldehyde
Hazard: Irritant
Formula:
C₉H₈N₂O |
MDL Number:
MFCD12962781 CAS Number: 1190321-17-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...