2,3-Dihydro-1H-pyrido[3,4-b][1,4]oxazine-8-carbaldehyde

Supplier: Matrix Scientific

054142-100MG 054142-500MG
101906-326EA 364.62 USD
101906-326 101906-328
2,3-Dihydro-1H-pyrido[3,4-b][1,4]oxazine-8-carbaldehyde
2,3-Dihydro-1H-pyrido[3,4-b][1,4]oxazine-8-carbaldehyde

Hazard: Irritant

Formula: C₈H₈N₂O₂
MDL Number: MFCD13563086
CAS Number: 1203499-38-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR