1-tert-Butyl-6-iodo-1H-pyrido[2,3-b][1,4]oxazin-2(3H)-one
Supplier: Matrix Scientific
054137-500MG
054137-100MG
101906-308EA
800
USD
101906-308
101906-306
1-tert-Butyl-6-iodo-1H-pyrido[2,3-b][1,4]oxazin-2(3H)-one
1-tert-Butyl-6-iodo-1H-pyrido[2,3-b][1,4]oxazin-2(3H)-one
Hazard: Irritant
Formula:
C₁₁H₁₃IN₂O₂ |
MDL Number:
MFCD13563081 CAS Number: 1203499-26-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...