6-Iodo-1H-pyrido[2,3-b][1,4]oxazin-2(3H)-one
Supplier: Matrix Scientific
054133-500MG
054133-100MG
101906-296EA
800
USD
101906-296
101906-294
6-Iodo-1H-pyrido[2,3-b][1,4]oxazin-2(3H)-one
6-Iodo-1H-pyrido[2,3-b][1,4]oxazin-2(3H)-one
Hazard: Irritant
Formula:
C₇H₅IN₂O₂ |
MDL Number:
MFCD13176674 CAS Number: 1203499-25-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...