tert-Butyl-5-((trimethylsilyl)ethynyl)-2,3-dihydro-1H-pyrido[3,4-b][1,4]oxazine-1-carboxylate
Supplier: Matrix Scientific
054130-100MG
054130-500MG
101906-284EA
800
USD
101906-284
101906-282
tert-Butyl-5-((trimethylsilyl)ethynyl)-2,3-dihydro-1H-pyrido[3,4-b][1,4]oxazine-1-carboxylate
tert-Butyl-5-((trimethylsilyl)ethynyl)-2,3-dihydro-1H-pyrido[3,4-b][1,4]oxazine-1-carboxylate
Hazard: Irritant
Formula:
C₁₇H₂₄N₂O₃Si MW: 332.48 g/mol |
MDL Number:
MFCD13176672 CAS Number: 1203499-24-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...