Methyl-1-(4-methoxybenzyl)-2-oxo-2,3-dihydro-1H-pyrido[2,3-b][1,4]oxazine-6-carboxylate

Supplier: Matrix Scientific

054129-100MG 054129-500MG
101906-278EA 364.62 USD
101906-278 101906-280
Methyl-1-(4-methoxybenzyl)-2-oxo-2,3-dihydro-1H-pyrido[2,3-b][1,4]oxazine-6-carboxylate
Methyl-1-(4-methoxybenzyl)-2-oxo-2,3-dihydro-1H-pyrido[2,3-b][1,4]oxazine-6-carboxylate

Hazard: Irritant

Formula: C₁₇H₁₆N₂O₅
MDL Number: MFCD13176671
CAS Number: 1203499-28-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR