6-(3-Hydroxyprop-1-ynyl)-1-(4-methoxybenzyl)-1H-pyrido[2,3-b][1,4]oxazin-2(3H)-one
Supplier: Matrix Scientific
054124-500MG
054124-100MG
101906-258EA
364.62
USD
101906-258
101906-260
6-(3-Hydroxyprop-1-ynyl)-1-(4-methoxybenzyl)-1H-pyrido[2,3-b][1,4]oxazin-2(3H)-one
6-(3-Hydroxyprop-1-ynyl)-1-(4-methoxybenzyl)-1H-pyrido[2,3-b][1,4]oxazin-2(3H)-one
Hazard: Irritant
Formula:
C₁₈H₁₆N₂O₄ |
MDL Number:
MFCD13176666 CAS Number: 1198425-67-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...