tert-Butyl-2-chloro-5-methylpyridin-3-ylcarbamate
Supplier: Matrix Scientific
054105-500MG
054105-100MG
101906-188EA
800
USD
101906-188
101906-186
tert-Butyl-2-chloro-5-methylpyridin-3-ylcarbamate
tert-Butyl-2-chloro-5-methylpyridin-3-ylcarbamate
Hazard: Irritant
Formula:
C₁₁H₁₅ClN₂O₂ MW: 242.71 g/mol |
MDL Number:
MFCD13563067 CAS Number: 1203498-98-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...