5-(3-((tert-Butyldimethylsilyloxy)methyl)pyrrolidin-1-yl)-2-chloro-3-iodopyridine

Supplier: Matrix Scientific

054103-100MG 054103-500MG
101906-178EA 364.62 USD
101906-178 101906-180
5-(3-((tert-Butyldimethylsilyloxy)methyl)pyrrolidin-1-yl)-2-chloro-3-iodopyridine
5-(3-((tert-Butyldimethylsilyloxy)methyl)pyrrolidin-1-yl)-2-chloro-3-iodopyridine

Hazard: Irritant

Formula: C₁₆H₂₆ClIN₂OSi
MW: 452.84 g/mol
MDL Number: MFCD13563065
CAS Number: 1203499-04-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR