1-(2-Methoxy-5-methylpyridin-3-yl)ethanone
Supplier: Matrix Scientific
054088-500MG
054088-100MG
101906-120EA
800
USD
101906-120
101906-118
1-(2-Methoxy-5-methylpyridin-3-yl)ethanone
1-(2-Methoxy-5-methylpyridin-3-yl)ethanone
Hazard: Irritant
Formula:
C₉H₁₁NO₂ |
MDL Number:
MFCD13563055 CAS Number: 1203499-64-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...