N-(3-(3-Hydroxypropyl)pyridin-2-yl)pivalamide
Supplier: Matrix Scientific
054071-100MG
054071-500MG
101906-052EA
800
USD
101906-052
101906-050
N-(3-(3-Hydroxypropyl)pyridin-2-yl)pivalamide
N-(3-(3-Hydroxypropyl)pyridin-2-yl)pivalamide
Hazard: Irritant
Formula:
C₁₃H₂₀N₂O₂ |
MDL Number:
MFCD13563043 CAS Number: 1203499-57-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...