2-Fluoro-3-iodo-6-(pyrrolidin-1-yl)pyridine
Supplier: Matrix Scientific
054062-500MG
054062-100MG
101906-016EA
800
USD
101906-016
101906-014
2-Fluoro-3-iodo-6-(pyrrolidin-1-yl)pyridine
2-Fluoro-3-iodo-6-(pyrrolidin-1-yl)pyridine
Hazard: Irritant
Formula:
C₉H₁₀FIN₂ |
MDL Number:
MFCD13176639 CAS Number: 1203499-32-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...