2-Chloro-3-iodo-5-methylpyridine

Supplier: Matrix Scientific

054060-100MG 054060-500MG
101906-006EA 364.62 USD
101906-006 101906-008
2-Chloro-3-iodo-5-methylpyridine
2-Chloro-3-iodo-5-methylpyridine

Hazard: Irritant

Formula: C₆H₅ClIN
MDL Number: MFCD09839273
CAS Number: 59782-91-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR