3-(3-(Benzyloxy)pyridin-2-yl)prop-2-yn-1-ol
Supplier: Matrix Scientific
054057-500MG
054057-100MG
101905-994EA
364.62
USD
101905-994
101905-996
3-(3-(Benzyloxy)pyridin-2-yl)prop-2-yn-1-ol
3-(3-(Benzyloxy)pyridin-2-yl)prop-2-yn-1-ol
Hazard: Irritant
Formula:
C₁₅H₁₃NO₂ |
MDL Number:
MFCD13176637 CAS Number: 1203499-09-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...