1-(5-Bromo-3-nitropyridin-2-yl)piperazine
Supplier: Matrix Scientific
054050-2.5G
054050-500MG
101905-966EA
380
USD
101905-966
101905-968
1-(5-Bromo-3-nitropyridin-2-yl)piperazine
1-(5-Bromo-3-nitropyridin-2-yl)piperazine
Hazard: Irritant
Formula:
C₉H₁₁BrN₄O₂ |
MDL Number:
MFCD13176633 CAS Number: 1203499-08-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...