2-(3-((tert-Butyldimethylsilyloxy)methyl)pyrrolidin-1-yl)-3-iodopyridine
Supplier: Matrix Scientific
054044-100MG
054044-500MG
101905-944EA
800
USD
101905-944
101905-942
2-(3-((tert-Butyldimethylsilyloxy)methyl)pyrrolidin-1-yl)-3-iodopyridine
2-(3-((tert-Butyldimethylsilyloxy)methyl)pyrrolidin-1-yl)-3-iodopyridine
Hazard: Irritant
Formula:
C₁₆H₂₇IN₂OSi |
MDL Number:
MFCD13176627 CAS Number: 1203499-01-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...