tert-Butyl-3-allyl-4-chloropyridin-2-ylcarbamate

Supplier: Matrix Scientific

054042-100MG 054042-500MG
101905-934EA 364.62 USD
101905-934 101905-936
tert-Butyl-3-allyl-4-chloropyridin-2-ylcarbamate
tert-Butyl-3-allyl-4-chloropyridin-2-ylcarbamate

Hazard: Irritant

Formula: C₁₃H₁₇ClN₂O₂
MW: 268.75 g/mol
MDL Number: MFCD13176625
CAS Number: 1203499-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR