1-Chloro-2-fluoro-3-(2,2,2-trifluoroethyl)benzene ≥97%

Supplier: Matrix Scientific

054027-1G
101905-888EA 238.46 USD
101905-888
1-Chloro-2-fluoro-3-(2,2,2-trifluoroethyl)benzene ≥97%
1-Chloro-2-fluoro-3-(2,2,2-trifluoroethyl)benzene

Hazard: Irritant

Formula: C₈H₅ClF₄
MDL Number: MFCD11226522
CAS Number: 1099598-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR