2-Methyl-2-(2-thienyl)propanoic acid

Supplier: Matrix Scientific

Synonyms: 2-Methyl-2-(thiophen-2-yl)propanoic acid

054008-500MG 054008-2.5G
101905-824EA 615.38 USD
101905-824 101905-826
2-Methyl-2-(2-thienyl)propanoic acid
2-Methyl-2-(2-thienyl)propanoic acid

Hazard: Irritant

Formula: C₈H₁₀O₂S
MW: 170.23 g/mol
MDL Number: MFCD13563034
CAS Number: 90953-78-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR