2-Methyl-2-(3-thienyl)propanoic acid

Supplier: Matrix Scientific

054007-500MG 054007-2.5G
101905-820EA 364.62 USD
101905-820 101905-822
2-Methyl-2-(3-thienyl)propanoic acid
2-Methyl-2-(3-thienyl)propanoic acid

Hazard: Irritant

Formula: C₈H₁₀O₂S
MDL Number: MFCD13563033
CAS Number: 147632-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR